missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Bromo-6-chlorobenzene-1,2,3,4-d4, TRC

Artikelnummer. 30335190
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30335190 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335190 Leverandør Toronto Research Chemicals Leverandørnr. TRCB682312250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Bromo-6-chlorobenzene-1,2,3,4-d4
CAS 1219795-51-3
Molekylær formel C6 D4 Br Cl
Mængde 250 mg
Synonym 2-Bromochlorobenzene-d4, Benzene-1,2,3,4-d4, 5-bromo-6-chloro- (ACI), 5-Bromo-6-chlorobenzene-1,2,3,4-d4 (ACI)
InChI formel InChI=1S/C6H4BrCl/c7-5-3-1-2-4-6(5)8/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(Br)c(Cl)c1[2H]
IUPAC navn 1-bromo-2-chloro-3,4,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 195.48
Formel vægt 193.9436
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.