missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-iodo-7-azaindole, 97%, Thermo Scientific™

Catalog Number p-7033036
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
757978-18-0
C7H4BrIN2
322.931
MFCD07779519
GIPGJYARDOQGDJ-UHFFFAOYSA-N
5-bromo-3-iodo-1h-pyrrolo 2,3-b pyridine, 5-bromo-3-iodo-7-azaindole, 1h-pyrrolo 2,3-b pyridine, 5-bromo-3-iodo, pubchem15707, endotherm enu336, ksc495s5j, 5-bromo-3-iodopyrrolo 2,3-b pyridine, 3-iodo-5-bromo-1h-pyrrolo 2,3-b-pyridine
24229212
5-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
C1=C2C(=CNC2=NC=C1Br)I
This item is not returnable. View return policy
757978-18-0
C7H4BrIN2
322.931
MFCD07779519
GIPGJYARDOQGDJ-UHFFFAOYSA-N
5-bromo-3-iodo-1h-pyrrolo 2,3-b pyridine, 5-bromo-3-iodo-7-azaindole, 1h-pyrrolo 2,3-b pyridine, 5-bromo-3-iodo, pubchem15707, endotherm enu336, ksc495s5j, 5-bromo-3-iodopyrrolo 2,3-b pyridine, 3-iodo-5-bromo-1h-pyrrolo 2,3-b-pyridine
24229212
5-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
C1=C2C(=CNC2=NC=C1Br)I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 322.93
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-iodo-7-azaindole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'