missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-fluoropyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4380055
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
669066-91-5
C6H3BrFNO2
220.00
MFCD07437865
JXHUXOAJAYWMHK-UHFFFAOYSA-N
21075872
5-bromo-3-fluoropyridine-2-carboxylic acid
OC(=O)C1=NC=C(Br)C=C1F
This item is not returnable. View return policy
669066-91-5
C6H3BrFNO2
220.00
MFCD07437865
JXHUXOAJAYWMHK-UHFFFAOYSA-N
21075872
5-bromo-3-fluoropyridine-2-carboxylic acid
OC(=O)C1=NC=C(Br)C=C1F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165°C to 170°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym 5-bromo-3-fluoropicolinic acid, 5-bromo-3-fluoro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-fluoro, 5-bromo-3-fluoropicolinicacid, 5-bromo-2-carboxy-3-fluoropyridine, 5-bromo-3-fluoropyridin-2-carboxylicacid, 2-pyridinecarboxylicacid,5-bromo-3-fluoro, 2-pyridinecarboxylicacid, 5-bromo-3-fluoro, 5-bromo-3-fluoro-pyridine-2-carboxylic acid, 5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid
Formula Weight 220
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-fluoropyridine-2-carboxylic acid
compliance-icons
Product Identifier
  • 5-Bromo-3-fluoropyridine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'