missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-fluoro-2-methoxypyridine, 97%, Thermo Scientific™

Catalog Number 15442079
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
124432-70-8
C6H5BrFNO
206.014
MFCD04112553
DEBFRAAJISQBIE-UHFFFAOYSA-N
14837011
5-bromo-3-fluoro-2-methoxypyridine
COC1=NC=C(C=C1F)Br
This item is not returnable. View return policy
124432-70-8
C6H5BrFNO
206.014
MFCD04112553
DEBFRAAJISQBIE-UHFFFAOYSA-N
14837011
5-bromo-3-fluoro-2-methoxypyridine
COC1=NC=C(C=C1F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 13638736
Synonym 2-methoxy-3-fluoro-5-bromopyridine, 5-bromo-3-fluoro-2-methoxy-pyridine, 5-bromo-3-fluoro-2-methoxypyrdine, pubchem10468, acmc-1c0iq, abbypharma ap-10-5273, pyridine,5-bromo-3-fluoro-2-methoxy, pyridine, 5-bromo-3-fluoro-2-methoxy
Formula Weight 206.01
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-3-fluoro-2-methoxypyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'