missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-3-chloro-2-fluoropyridine, 96%

Brand:  Alfa Aesar™ H64547.MD

Additional Details : CAS Number : 38185-56-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15471819

  • 203.60 DKK / 250mg


Estimated Shipment date
from Supplier 03-07-2020
Add to basket

Specifications

Chemical Identifiers

38185-56-7
210.43
WXKLHBHJIBAXQH-UHFFFAOYSA-N
12056904
C1=C(C(=NC=C1Br)F)Cl
C5H2BrClFN
MFCD09834365
2-fluoro-3-chloro-5-bromopyridine, 3-bromo-5-chloro-6-fluoropyridine, 5-bromo-3-chloro-2-fluoro-pyridine, acmc-1ct80, 5-bromo-3-chloro-2-fluoropyridine,, pyridine,5-bromo-3-chloro-2-fluoro, pyridine, 5-bromo-3-chloro-2-fluoro
5-bromo-3-chloro-2-fluoropyridine

Specifications

5-Bromo-3-chloro-2-fluoropyridine
250mg
C5H2BrClFN
1563886
WXKLHBHJIBAXQH-UHFFFAOYSA-N
5-bromo-3-chloro-2-fluoropyridine
12056904
Solid
60°C
38185-56-7
MFCD09834365
2-fluoro-3-chloro-5-bromopyridine, 3-bromo-5-chloro-6-fluoropyridine, 5-bromo-3-chloro-2-fluoro-pyridine, acmc-1ct80, 5-bromo-3-chloro-2-fluoropyridine,, pyridine,5-bromo-3-chloro-2-fluoro, pyridine, 5-bromo-3-chloro-2-fluoro
C1=C(C(=NC=C1Br)F)Cl
210.43
210.43
96%
Videos
SDS Documents