missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyridine-3-carboxaldehyde, 99%, Thermo Scientific™

Catalog Number p-7033053
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
103058-87-3
C7H6BrNO2
216.03
MFCD05663521
VRNOWEKCASDTFG-UHFFFAOYSA-N
5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, pubchem9207, abbypharma ap-10-5904, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
1516403
5-bromo-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(Br)C=C1C=O
This item is not returnable. View return policy
103058-87-3
C7H6BrNO2
216.03
MFCD05663521
VRNOWEKCASDTFG-UHFFFAOYSA-N
5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, pubchem9207, abbypharma ap-10-5904, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
1516403
5-bromo-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(Br)C=C1C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-methoxypyridine-3-carboxaldehyde is used as a synthetic building block. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 216.04
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-methoxypyridine-3-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'