missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dichloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7028195
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
36082-50-5
C4HBrCl2N2
227.87
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine, 2,4-dichloro-5-bromo pyrimidine, 5-bromo-2,4-dichloro-pyrimidine, pyrimidine, 5-bromo-2,4-dichloro, 5-bromo-2,6-dichloropyrimidine, 5-bromo-2,4-dichloro-1,3-diazine, 5-brom-2,4-dichlorpyrimidin, zlchem 380, pubchem3993, acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br
This item is not returnable. View return policy
36082-50-5
C4HBrCl2N2
227.87
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine, 2,4-dichloro-5-bromo pyrimidine, 5-bromo-2,4-dichloro-pyrimidine, pyrimidine, 5-bromo-2,4-dichloro, 5-bromo-2,6-dichloropyrimidine, 5-bromo-2,4-dichloro-1,3-diazine, 5-brom-2,4-dichlorpyrimidin, zlchem 380, pubchem3993, acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29°C to 30°C
Density 1.781
Boiling Point 128°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.603
Quantity 1 g
UN Number UN2923
Beilstein 124441
Formula Weight 227.88
Percent Purity 98%
Chemical Name or Material 5-Bromo-2,4-dichloropyrimidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'