missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methyl-1H-indole, 97%, Thermo Scientific™

Catalog Number p-4510635
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
10075-52-2
C9H8BrN
210.07
MFCD00159857
SBOITLSQLQGSLO-UHFFFAOYSA-N
285757
5-bromo-1-methylindole
CN1C=CC2=CC(Br)=CC=C12
This item is not returnable. View return policy
10075-52-2
C9H8BrN
210.07
MFCD00159857
SBOITLSQLQGSLO-UHFFFAOYSA-N
285757
5-bromo-1-methylindole
CN1C=CC2=CC(Br)=CC=C12
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-bromo-1-methyl-1h-indole, 5-bromo-1-methyl-indole, 1h-indole, 5-bromo-1-methyl, 1-methyl-5-bromoindole, 1h-indole,5-bromo-1-methyl, pubchem7946, zlchem 1345, n-methyl-5-bromoindole, 5-bromo-n-methylindole, acmc-2097se
Formula Weight 210.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-methyl-1H-indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'