missing translation for 'onlineSavingsMsg'
Learn More

5-bromo-1-methyl-1H-indole-3-carbaldehyde, Thermo Scientific™

Catalog Number p-4391214
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
10102-94-0
C10H8BrNO
238.084
NZJJLQUTQOICBN-UHFFFAOYSA-N
5-bromo-1-methyl-1h-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde,5-bromo-1-methyl, 1h-indole-3-carboxaldehyde, 5-bromo-1-methyl, chembrdg-bb 5100225, 1-methyl-5-bromoindole-3-aldehyde, 1-methyl-5-bromo-1h-indole-3-carbaldehyde
778451
5-bromo-1-methylindole-3-carbaldehyde
CN1C=C(C2=C1C=CC(=C2)Br)C=O
This item is not returnable. View return policy
10102-94-0
C10H8BrNO
238.084
NZJJLQUTQOICBN-UHFFFAOYSA-N
5-bromo-1-methyl-1h-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde,5-bromo-1-methyl, 1h-indole-3-carboxaldehyde, 5-bromo-1-methyl, chembrdg-bb 5100225, 1-methyl-5-bromoindole-3-aldehyde, 1-methyl-5-bromo-1h-indole-3-carbaldehyde
778451
5-bromo-1-methylindole-3-carbaldehyde
CN1C=C(C2=C1C=CC(=C2)Br)C=O
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.08
Percent Purity 97%
Chemical Name or Material 5-bromo-1-methyl-1H-indole-3-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'