Få mere at vide
5-Benzyloxytryptophol-alpha,alpha,beta,beta-d4, CDN
Artikelnummer. 30325256
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325256 0.05 g
30311019 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325256 Leverandør CDN Leverandørnr. CDND72860.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(Phenylmethoxy)-1 H-indole-3-ethan-alpha,alpha,beta,beta-d4-ol
CAS 75238-47-0
Molekylær formel C17 D4 H13 N O2
Mængde 0.05 g
Synonym 1 H-Indole-3-ethan-α,α,β,β-d4-ol, 5-(phenylmethoxy)- (9 CI), 5-(Phenylmethoxy)-1 H-indole-3-ethan-α,α,β,β-d4-ol, 2-(5-Benzyloxy-1 H-indol-3-yl)-ethanol, 5-Benzyloxytryptophol-α,α,β,β-d4, 5-Benzyloxy-1 H-indole-3-ethanol-D4 (ethanol-D4)
InChI formel InChI=1 S/C17H17NO2/c19-9-8-14-11-18-17-7-6-15(10-16(14)17)20-12-13-4-2-1-3-5-13/h1-7,10-11,18-19 H,8-9,12H2/i8D2,9D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])c1c[nH]c2ccc(OCc3ccccc3)cc12
IUPAC navn 1,1,2,2-tetradeuterio-2-(5-phenylmethoxy-1 H-indol-3-yl)ethanol
Molekylvægt (g/mol) 271.347
Formel vægt 271.151 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity