Learn More
5-(Benzyloxycarbonylamino)valeric acid, 97%
CAS: 23135-50-4 | C13H17NO4 | 251.28 g/mol
90.48 DKK - 1034.46 DKK
Chemical Identifiers
CAS | 23135-50-4 |
---|---|
Molecular Formula | C13H17NO4 |
Molecular Weight (g/mol) | 251.28 |
MDL Number | MFCD00188458 |
InChI Key | QYYPKLYDFCYGPG-UHFFFAOYSA-N |
Synonym | 5-benzyloxy carbonyl amino pentanoic acid, 5-benzyloxycarbonylamino-pentanoic acid, 5-benzyloxycarbonylamino pentanoic acid, z-5-ava-oh, 5-carbobenzoxyamino valeric acid, 5-carbobenzoxyamino pentanoic acid, 5-phenylmethoxycarbonylamino pentanoic acid, 5-benzyloxycarbonylamino-pentanoicacid, pubchem20738 |
PubChem CID | 3690024 |
SMILES | OC(=O)CCCCNC(=O)OCC1=CC=CC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23135-50-4 | |
251.28 | |
QYYPKLYDFCYGPG-UHFFFAOYSA-N | |
3690024 |
C13H17NO4 | |
MFCD00188458 | |
5-benzyloxy carbonyl amino pentanoic acid, 5-benzyloxycarbonylamino-pentanoic acid, 5-benzyloxycarbonylamino pentanoic acid, z-5-ava-oh, 5-carbobenzoxyamino valeric acid, 5-carbobenzoxyamino pentanoic acid, 5-phenylmethoxycarbonylamino pentanoic acid, 5-benzyloxycarbonylamino-pentanoicacid, pubchem20738 | |
OC(=O)CCCCNC(=O)OCC1=CC=CC=C1 |
Specifications
99°C to 105°C | |
White | |
C6H4CH2OCONH2(CH2)4CO2H | |
2334766 | |
QYYPKLYDFCYGPG-UHFFFAOYSA-N | |
5-{[(benzyloxy)carbonyl]amino}pentanoic acid | |
3690024 | |
97% | |
5-(Benzyloxycarbonylamino)valeric acid |
23135-50-4 | |
C13H17NO4 | |
MFCD00188458 | |
5-benzyloxy carbonyl amino pentanoic acid, 5-benzyloxycarbonylamino-pentanoic acid, 5-benzyloxycarbonylamino pentanoic acid, z-5-ava-oh, 5-carbobenzoxyamino valeric acid, 5-carbobenzoxyamino pentanoic acid, 5-phenylmethoxycarbonylamino pentanoic acid, 5-benzyloxycarbonylamino-pentanoicacid, pubchem20738 | |
OC(=O)CCCCNC(=O)OCC1=CC=CC=C1 | |
251.28 | |
251.28 | |
Solid |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.