Få mere at vide
5-Benzyloxy Rosiglitazone-d4, TRC
Artikelnummer. 30345929
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30345929 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345929 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28816210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Benzyloxy Rosiglitazone-d4
CAS 1794768-28-7
Molekylær formel C25H21D4N3O4S
Mængde 10 mg
Synonym 5-{4-[2-[(5-Benzyloxypyridin-2-yl)methylamino]ethoxy]benzyl}thiazolidine-2,4-dione-d4, 5-[[3-[2-[Methyl[5-(phenylmethoxy)-2-pyridinyl]amino]ethoxy]phenyl]methyl]-2,4-thiazolidinedione-d4
InChI formel InChI=1S/C25H25N3O4S/c1-28(23-12-11-21(16-26-23)32-17-19-5-3-2-4-6-19)13-14-31-20-9-7-18(8-10-20)15-22-24(29)27-25(30)33-22/h2-12,16,22H,13-15,17H2,1H3,(H,27,29,30)/i13D2,14D2
SMIL [2H]C([2H])(Oc1ccc(CC2SC(=O)NC2=O)cc1)C([2H])([2H])N(C)c3ccc(OCc4ccccc4)cn3
IUPAC navn 5-[[4-[1,1,2,2-tetradeuterio-2-[methyl-(5-phenylmethoxypyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
Molekylvægt (g/mol) 467.57
Formel vægt 467.1817