Få mere at vide
5-Benzyloxy Rosiglitazone-d4, TRC
Artikelnummer. 30331160
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30331160 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331160 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2881621MG
Denne vare kan ikke returneres. Se returpolitik
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Benzyloxy Rosiglitazone-d4
CAS 1794768-28-7
Molekylær formel C25H21D4N3O4S
Mængde 1 mg
Synonym 5-{4-[2-[(5-Benzyloxypyridin-2-yl)methylamino]ethoxy]benzyl}thiazolidine-2,4-dione-d4, 5-[[3-[2-[Methyl[5-(phenylmethoxy)-2-pyridinyl]amino]ethoxy]phenyl]methyl]-2,4-thiazolidinedione-d4
InChI formel InChI=1S/C25H25N3O4S/c1-28(23-12-11-21(16-26-23)32-17-19-5-3-2-4-6-19)13-14-31-20-9-7-18(8-10-20)15-22-24(29)27-25(30)33-22/h2-12,16,22H,13-15,17H2,1H3,(H,27,29,30)/i13D2,14D2
SMIL [2H]C([2H])(Oc1ccc(CC2SC(=O)NC2=O)cc1)C([2H])([2H])N(C)c3ccc(OCc4ccccc4)cn3
IUPAC navn 5-[[4-[1,1,2,2-tetradeuterio-2-[methyl-(5-phenylmethoxypyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
Molekylvægt (g/mol) 467.57
Formel vægt 467.1817