Få mere at vide
5-Benzyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride, Mikromol™
Beskrivelse
5-Benzyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 5-Benzyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride |
| CAS | 54903-07-0 |
| Analyt- eller komponentnavne | 5-Benzyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H15 N S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Ticlopidine Hydrochloride Imp. D (EP), Ticlopidine Imp. D (EP), 5-Benzyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride, Ticlopidine Hydrochloride Imp. D (EP) as Hydrochloride, Ticlopidine Hydrochloride Impurity D as Hydrochloride, Ticlopidine Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C14H15NS.ClH/c1-2-4-12(5-3-1)10-15-8-6-14-13(11-15)7-9-16-14;/h1-5,7,9H,6,8,10-11H2;1H |
| SMIL | Cl.C(N1CCc2sccc2C1)c3ccccc3 |
| IUPAC navn | 5-benzyl-6,7-dihydro-4H-thieno[3,2-c]pyridine;hydrochloride |
| Vis mere |