Få mere at vide
∆5-Androstene-3b,17b-diol-16,16,17-d3, TRC
Artikelnummer. 30345462
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30345462 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345462 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63754725MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Androsten-3beta,17beta-diol-D3
CAS 73609-45-7
Molekylær formel C19 2H3 H27 O2
Mængde 25 mg
Synonym Androst-5-ene-16,16,17-d3-3,17-diol, (3β,17β)- (9CI), 5-Androsten-3β,17β-diol-d3, 5-Androsten-3β,17β-diol-16,16,17-D3, ∆5-Androstene-3β,17β-diol-16,16,17-d3, (3β,17β)-Androst-5-ene-16,16,17-d3-3,17-diol
InChI formel InChI=1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-17,20-21H,4-11H2,1-2H3/t13-,14-,15-,16-,17-,18-,19-/m0/s1/i6D2,17D
SMIL [2H]C1([2H])C[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1([2H])O
IUPAC navn (3S,8R,9S,10R,13S,14S,17S)-16,16,17-trideuterio-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 293.46
Formel vægt 293.2434