missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Aminopentanenitrile Hydrochloride, TRC

Artikelnummer. 30327890
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327890 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30327890 Leverandør Toronto Research Chemicals Leverandørnr. TRCA629958100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Aminopentanenitrile Hydrochloride
CAS 1638108-03-8
Molekylær formel C5H10N2 . HCl
Mængde 100 mg
InChI formel InChI=1S/C5H10N2/c6-4-2-1-3-5-7/h1-4,6H2
SMIL NCCCCC#N
IUPAC navn 5-aminopentanenitrile
Molekylvægt (g/mol) 134.61
Formel vægt 98.0844
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.