Få mere at vide
5-Aminolevulinic Acid Methyl Ester Hydrochloride, TRC
Artikelnummer. 30133244
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133244 10 g
30006418 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133244 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63258510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79416-27-6
Molekylær formel C6 H11 N O3 . HCl
Molekylvægt (g/mol) 181.62
Synonym Pentanoic acid, 5-amino-4-oxo-, methyl ester, hydrochloride (1:1), Pentanoic acid, 5-amino-4-oxo-, methyl ester, hydrochloride (9CI), 5-Amino-4-oxopentanoic acid methyl ester hydrochloride, 5-Aminolevulinic acid methyl ester hydrochloride, Methyl 5-amino-4-oxopentanoate hydrochloride, Methyl 5-aminolevulinate hydrochloride, Methyl ALA ester hydrochloride, Methyl aminolevulinate hydrochloride, Metvix, P 1202, delta-Aminolevulinic acid methyl ester hydrochloride, 5-Amino-4-oxopentanoic Acid Methyl Ester Hydrochloride (1:1), Methyl 5-Amino-4-oxopentanoate Hydrochloride, Methyl 5-Aminolevulinate Hydrochloride, Methyl ALA Ester Hydrochloride, Methyl Aminolevulinate Hydrochloride, Metvix, P 1202, δ-Aminolevulinic Acid Methyl Ester Hydrochloride;
IUPAC navn methyl 5-amino-4-oxopentanoate hydrochloride
SMIL Cl.O=C(OC)CCC(=O)CN
Tekniske data
Kemisk navn eller materiale 5-Aminolevulinic Acid Methyl Ester Hydrochloride
InChI formel InChI=1S/C6H11NO3.ClH/c1-10-6(9)3-2-5(8)4-7;/h2-4,7H2,1H3;1H
Mængde 10 g
Formel vægt 181.0506