Få mere at vide
5-Aminoimidazole-4-carboxamide-1-Beta-D-Ribofuranosyl 5’-Monophosphate, TRC
Artikelnummer. 30005853
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005853 50 mg
30003279 5 mg
30003385 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005853 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61170550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3031-94-5
Molekylær formel C9 H15 N4 O8 P
Molekylvægt (g/mol) 338.21
IUPAC navn [(2R,3S,4R,5R)-5-(4-amino-5-carbamoylimidazol-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMIL NC(=O)c1c(N)ncn1[C@@H]2O[C@H](COP(=O)(O)O)[C@@H](O)[C@H]2O
Tekniske data
Kemisk navn eller materiale 5-Aminoimidazole-4-carboxamide-1-beta-D-Ribofuranosyl 5'-Monophosphate
Analyt- eller komponentnavne 5-Aminoimidazole-4-carboxamide-1-beta-D-Ribofuranosyl 5'-Monophosphate
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C9H15N4O8P/c10-7-4(8(11)16)13(2-12-7)9-6(15)5(14)3(21-9)1-20-22(17,18)19/h2-3,5-6,9,14-15H,1,10H2,(H2,11,16)(H2,17,18,19)/t3-,5-,6-,9-/m1/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 50 mg
Formel vægt 338.0627
Procent renhed >95%