Få mere at vide
5-Aminoimidazole-4-carboxamide-1-beta-D-Ribofuranosyl 5’-Monophosphate Disodium Salt, TRC
Artikelnummer. 30327089
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327089 25 mg
30326989 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30327089 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61258525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Aminoimidazole-4-carboxamide-1-beta-D-Ribofuranosyl 5'-Monophosphate Disodium Salt
CAS 6884-89-5
Molekylær formel C9 H13 N4 Na2 O8 P
Mængde 25 mg
Synonym 5(4)-Amino-4(5)-imidazolecarboxamide Riboside 5'-Phosphoric Acid Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(5-Amino-4-carbamoyl-1H-imidazol-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl Phosphate
InChI formel InChI=1S/C9H15N4O8P.2Na/c10-7-4(8(11)16)12-2-13(7)9-6(15)5(14)3(21-9)1-20-22(17,18)19;;/h2-3,5-6,9,14-15H,1,10H2,(H2,11,16)(H2,17,18,19);;/q;2*+1/p-2/t3-,5-,6-,9-;;/m1../s1
SMIL NC(=O)c1ncn([C@@H]2O[C@H](COP(=O)(O[Na])O[Na])[C@@H](O)[C@H]2O)c1N
IUPAC navn [[(2R,3S,4R,5R)-5-(5-amino-4-carbamoylimidazol-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-sodiooxyphosphoryl]oxysodium
Molekylvægt (g/mol) 382.175
Formel vægt 382.027