missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
200 mg
Description
5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside is used as a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside is used as a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes.
Solubility
Soluble in DMSO at 2mg/ml. Soluble in water or ethanol at less than 1mg/ml
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside is used as a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes.
Solubility
Soluble in DMSO at 2mg/ml. Soluble in water or ethanol at less than 1mg/ml
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | 214°C to 215°C |
Color | Brown |
Quantity | 50 mg |
Merck Index | 14,16 |
Solubility Information | Soluble in DMSO at 2mg/ml; Soluble in water or ethanol at less than 1mg/ml |
Formula Weight | 258.23 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside |
Safety and Handling
Product Identifier
- 5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 5-amino-1-.beta.-D-ribofuranosyl-1H-Imidazole-4-carboxamide
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'