missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Amino-7-azaindole, 97%

Brand:  Alfa Aesar™ H35382.03

Additional Details : CAS Number : 100960-07-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487507

  • 4154.08 DKK / 1g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

100960-07-4
133.154
PLWBENCHEUFMTN-UHFFFAOYSA-N
11715187
C1=CNC2=NC=C(C=C21)N
C7H7N3
MFCD06659683
5-amino-7-azaindole, 1h-pyrrolo 2,3-b pyridin-5-amine, 1h-pyrrolo 2,3-b pyridin-5-ylamine, 1h-pyrrolo 2,3-b pyridine-5-amine, 5-amino-1h-pyrrolo 2,3-b pyridine, 5-amino-7-aza-1h-indole, pubchem15250, acmc-2097tg, pyrrolo 2,3-b pyrid-5-ylamine, pyrrolo 2,3-b pyridin-5-yl amine
1H-pyrrolo[2,3-b]pyridin-5-amine

Specifications

5-Amino-7-azaindole
Brown-Yellow to Yellow
1g
C7H7N3
5-amino-7-azaindole, 1h-pyrrolo 2,3-b pyridin-5-amine, 1h-pyrrolo 2,3-b pyridin-5-ylamine, 1h-pyrrolo 2,3-b pyridine-5-amine, 5-amino-1h-pyrrolo 2,3-b pyridine, 5-amino-7-aza-1h-indole, pubchem15250, acmc-2097tg, pyrrolo 2,3-b pyrid-5-ylamine, pyrrolo 2,3-b pyridin-5-yl amine
C1=CNC2=NC=C(C=C21)N
133.154
133.15
97%
Light sensitive
129°C to 131°C
100960-07-4
MFCD06659683
PLWBENCHEUFMTN-UHFFFAOYSA-N
1H-pyrrolo[2,3-b]pyridin-5-amine
11715187
Solid
Videos
SDS Documents