Få mere at vide
5-Amino-2-nitrobenzophenone-d5, TRC
Artikelnummer. 30314528
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30314528 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314528 Leverandør Toronto Research Chemicals Leverandørnr. TRCA618402100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Amino-2-nitrobenzophenone-d5
CAS 1189718-61-3
Molekylær formel C13H5D5N2O3
Mængde 100 mg
Synonym (2-Amino-5-nitrophenyl)(phenyl-d5)methanone, 5-Nitro-2-aminobenzophenone-d5, Ro 7-19990-d5
InChI formel InChI=1S/C13H10N2O3/c14-12-7-6-10(15(17)18)8-11(12)13(16)9-4-2-1-3-5-9/h1-8H,14H2/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(=O)c2cc(ccc2N)[N+](=O)[O-])c([2H])c1[2H]
IUPAC navn (2-amino-5-nitrophenyl)-(2,3,4,5,6-pentadeuteriophenyl)methanone
Molekylvægt (g/mol) 247.26