missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methoxypyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7030002
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
6628-77-9
C6H8N2O
124.143
MFCD00006264
UUVDJIWRSIJEBS-UHFFFAOYSA-N
5-amino-2-methoxypyridine, 2-methoxy-5-aminopyridine, 6-methoxy-3-pyridylamine, 3-pyridinamine, 6-methoxy, 2-methoxy-5-amino pyridine, 6-methoxy-pyridin-3-ylamine, pyridine, 5-amino-2-methoxy, 6-methoxy-3-pyridinylamine, 6-methoxy-3-pyridinamine
81121
6-methoxypyridin-3-amine
COC1=NC=C(C=C1)N
This item is not returnable. View return policy
6628-77-9
C6H8N2O
124.143
MFCD00006264
UUVDJIWRSIJEBS-UHFFFAOYSA-N
5-amino-2-methoxypyridine, 2-methoxy-5-aminopyridine, 6-methoxy-3-pyridylamine, 3-pyridinamine, 6-methoxy, 2-methoxy-5-amino pyridine, 6-methoxy-pyridin-3-ylamine, pyridine, 5-amino-2-methoxy, 6-methoxy-3-pyridinylamine, 6-methoxy-3-pyridinamine
81121
6-methoxypyridin-3-amine
COC1=NC=C(C=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29°C to 31°C
Boiling Point 85°C to 90°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5735
Quantity 5 g
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material 5-Amino-2-methoxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'