missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-hydroxy-4'-sulfo[1,1'-biphenyl]-3-carboxylic Acid, TRC

Product Code. 30322433
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30322433 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322433 Supplier Toronto Research Chemicals Supplier No. TRCA6294105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-(4-Sulfophenyl)-5-aminosalicylic acid
CAS 887256-40-8
Molekylær formel C13 H11 N O6 S
Mængde 5 mg
Synonym 5-Amino-2-hydroxy-4'-sulfo[1,1'-biphenyl]-3-carboxylic Acid, 5-Amino-2-hydroxy-3-(4-sulfophenyl)benzoic Acid, 3-(4-Sulfophenyl)-5-aminosalicylic Acid, Mesalamine impurity P, 3-(4-sulfophenyl)-5-aminosalicylic Acid
InChI formel InChI=1S/C13H11NO6S/c14-8-5-10(12(15)11(6-8)13(16)17)7-1-3-9(4-2-7)21(18,19)20/h1-6,15H,14H2,(H,16,17)(H,18,19,20)
SMIL Nc1cc(C(=O)O)c(O)c(c1)c2ccc(cc2)S(=O)(=O)O
IUPAC navn 5-amino-2-hydroxy-3-(4-sulfophenyl)benzoic acid
Molekylvægt (g/mol) 309.29
Formel vægt 309.0307
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.