missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-carboxy-4-cyano-3-thiopheneacetic Acid, TRC

Product Code. 30323753
Change view
Click to view available options
Mængde:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30323753 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323753 Supplier Toronto Research Chemicals Supplier No. TRCA6041502.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Amino-2-carboxy-4-cyano-3-thiopheneacetic Acid
CAS 674773-14-9
Molekylær formel C8H6N2O4S
Mængde 2.5 mg
Synonym 5-Amino-3-(carboxymethyl)-4-cyanothiophene-2-carboxylic Acid
InChI formel InChI=1S/C8H6N2O4S/c9-2-4-3(1-5(11)12)6(8(13)14)15-7(4)10/h1,10H2,(H,11,12)(H,13,14)
SMIL O=C(O)CC1=C(C(O)=O)SC(N)=C1C#N
IUPAC navn 5-amino-3-(carboxymethyl)-4-cyanothiophene-2-carboxylic acid
Molekylvægt (g/mol) 226.21
Formel vægt 226
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.