missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Amino-2,4-dichloropyridine, 97%

Brand:  Alfa Aesar™ H66282.03

Additional Details : CAS Number : 7321-93-9 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15492719

  • 1290.00 DKK / 1g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

7321-93-9
163.001
FBGVTWONYOCYGA-UHFFFAOYSA-N
595729
C1=C(C(=CN=C1Cl)N)Cl
C5H4Cl2N2
MFCD00234054
5-amino-2,4-dichloropyridine, 4,6-dichloro-pyridin-3-ylamine, 3-pyridinamine, 4,6-dichloro, 4,6-dichloro-3-pyridinamine, 4,6-dichloropyridin-3-ylamine, pubchem17045, acmc-209opx, 2,4-dichloro-5-aminopyridine, 4,6-dichloropyridine-3-amine, 3-pyridinamine,4,6-dichloro
4,6-dichloropyridin-3-amine

Specifications

5-Amino-2,4-dichloropyridine
1g
C5H4Cl2N2
UN2811
5-amino-2,4-dichloropyridine, 4,6-dichloro-pyridin-3-ylamine, 3-pyridinamine, 4,6-dichloro, 4,6-dichloro-3-pyridinamine, 4,6-dichloropyridin-3-ylamine, pubchem17045, acmc-209opx, 2,4-dichloro-5-aminopyridine, 4,6-dichloropyridine-3-amine, 3-pyridinamine,4,6-dichloro
C1=C(C(=CN=C1Cl)N)Cl
163.001
163
97%
Brown
7321-93-9
MFCD00234054
119452
FBGVTWONYOCYGA-UHFFFAOYSA-N
4,6-dichloropyridin-3-amine
595729
Powder
Videos
SDS Documents