missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(2,4-dichlorophenoxy)pyridine, 99%, Thermo Scientific™

Catalog Number 11470230
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
85686-48-2
C11H8Cl2N2O
255.10
MFCD00052643
XBBAKFQNAVTALV-UHFFFAOYSA-N
6-2,4-dichlorophenoxy pyridin-3-amine, 5-amino-2-2,4-dichlorophenoxy pyridine, 6-2,4-dichlorophenoxy-3-pyridylamine, acmc-20am6j, maybridge1_001016, 2,4-dichlorophenoxy-5-pyridinamine, 6-2,4-dichloro-phenoxy-pyridin-3-ylamine, 6-2,4-dichlorophenoxy pyridin-3-amine,97
735776
6-(2,4-dichlorophenoxy)pyridin-3-amine
NC1=CN=C(OC2=C(Cl)C=C(Cl)C=C2)C=C1
This item is not returnable. View return policy
85686-48-2
C11H8Cl2N2O
255.10
MFCD00052643
XBBAKFQNAVTALV-UHFFFAOYSA-N
6-2,4-dichlorophenoxy pyridin-3-amine, 5-amino-2-2,4-dichlorophenoxy pyridine, 6-2,4-dichlorophenoxy-3-pyridylamine, acmc-20am6j, maybridge1_001016, 2,4-dichlorophenoxy-5-pyridinamine, 6-2,4-dichloro-phenoxy-pyridin-3-ylamine, 6-2,4-dichlorophenoxy pyridin-3-amine,97
735776
6-(2,4-dichlorophenoxy)pyridin-3-amine
NC1=CN=C(OC2=C(Cl)C=C(Cl)C=C2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 255.11
Percent Purity 99%
Chemical Name or Material 5-Amino-2-(2,4-dichlorophenoxy)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'