Få mere at vide
5-Amino-1,3,4-thiadiazole-2-sulfonamide, Mikromol™
Artikelnummer. 30050370
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050370 25 mg
30049500 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050370 Leverandør Mikromol Leverandørnr. MM0648.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Amino-1,3,4-thiadiazole-2-sulfonamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Amino-1,3,4-thiadiazole-2-sulfonamide
CAS 14949-00-9
Analyt- eller komponentnavne 5-Amino-1,3,4-thiadiazole-2-sulfonamide
Forsendelsestilstand Room Temperature
Molekylær formel C2 H4 N4 O2 S2
Certificeringer/compliance ISO 17025
Synonym Acetazolamide Related Compound D, Acetazolamide Imp. D (EP), Acetazolamide USP Related Compound D, Acetazolamide USP RC D, 1,3,4-Thiadiazole-2-sulfonamide, 5-amino-, 2-Amino-1,3,4-thiadiazole-5-sulfonamide, 5-Amino-1,3,4-thiadiazol-2-sulfonamide, 5-Amino-1,3,4-thiadiazole-2-sulfonamide, CL 5343, NSC 22979, Tio-urasin, Acetazolamide Related Compound D, Acetazolamide Impurity D
InChI formel InChI=1S/C2H4N4O2S2/c3-1-5-6-2(9-1)10(4,7)8/h(H2,3,5)(H2,4,7,8)
SMIL Nc1nnc(s1)S(=O)(=O)N
IUPAC navn 5-amino-1,3,4-thiadiazole-2-sulfonamide
Molekylvægt (g/mol) 180.21
Mængde 25 mg
Formel vægt 179.9776 g/mol