missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylindane, 97%, Thermo Scientific Chemicals

Catalog Number p-7024443
Click to view available options
Quantity:
2.5 g
10 g
50 g
This item is not returnable. View return policy
4228-10-8
MFCD00021246
This item is not returnable. View return policy
4228-10-8
MFCD00021246

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 85°C to 87°C (0.05 mmHg)
Molecular Formula C11H12O
Quantity 2.5 g
Synonym 5-acetylindane, 5-acetylindan, 1-2,3-dihydro-1h-inden-5-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl ethan-1-one, ethanone,3-dihydro-1h-inden-5-yl, indan-5-ylethan-1-one, 1-indane-5-yl-ethanone, acmc-1arc8, 1-2,3-dihydro-1h-inden-6-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl-ethanone
InChI Key HCMWPTFPUCPKQM-UHFFFAOYSA-N
SMILES CC(=O)C1=CC2=C(CCC2)C=C1
IUPAC Name 1-(2,3-dihydro-1H-inden-5-yl)ethanone
Molecular Weight (g/mol) 160.22
PubChem CID 233976
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material 5-Acetylindane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'