missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Acetylamino-6-amino-3-methyluracil Hydrate, TRC

Artikelnummer. 30319200
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319200 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30319200 Leverandør Toronto Research Chemicals Leverandørnr. TRCA168125100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Acetylamino-6-amino-3-methyluracil, Hydrate
CAS 1196153-01-1
Molekylær formel C7H12N4O4
Mængde 100 mg
Synonym Acetamide, N-(4-amino-1,2,3,6-tetrahydro-1-methyl-2,6-dioxo-5-pyrimidinyl)-, hydrate (1:1), N-(4-Amino-1,2,3,6-tetrahydro-1-methyl-2,6-dioxo-5-pyrimidinyl)acetamide monohydrate, 5-Acetylamino-6-amino-3-methyluracil monohydrate, 5-Acetamido-6-amino-3-methyluracil monohydrate
InChI formel InChI=1S/C7H10N4O3.H2O/c1-3(12)9-4-5(8)10-7(14)11(2)6(4)13;/h8H2,1-2H3,(H,9,12)(H,10,14);1H2
SMIL O.CN1C(=O)NC(=C(NC(=O)C)C1=O)N
IUPAC navn N-(6-amino-3-methyl-2,4-dioxo-1H-pyrimidin-5-yl)acetamide;hydrate
Molekylvægt (g/mol) 216.19
Formel vægt 216.0859
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.