missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Acetyl-8-(phenylmethoxy)-2-quinolinone, TRC

Artikelnummer. 30324432
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30324432 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324432 Supplier Toronto Research Chemicals Supplier No. TRCA187530100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Acetyl-8-(phenylmethoxy)-2-quinolinone
CAS 93609-84-8
Molekylær formel C18H15NO3
Mængde 100 mg
Synonym 5-Acetyl-8-(benzyloxy)carbostyril, 5-Acetyl-8-benzyloxy-1H-quinolin-2-one, 5-Acetyl-8-benzyloxy-2(1H)-quinolinone, 8-(Benzyloxy)-5-(1-oxoethyl)-1H-quinolin-2-one
InChI formel InChI=1S/C18H15NO3/c1-12(20)14-7-9-16(18-15(14)8-10-17(21)19-18)22-11-13-5-3-2-4-6-13/h2-10H,11H2,1H3,(H,19,21)
SMIL CC(=O)c1ccc(OCc2ccccc2)c3NC(=O)C=Cc13
IUPAC navn 5-acetyl-8-phenylmethoxy-1H-quinolin-2-one
Molekylvægt (g/mol) 293.32
Formel vægt 293.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.