missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Acetyl-2H-pyrazole-3-carboxylic Acid Ethyl Ester, TRC

Artikelnummer. 30319588
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319588 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30319588 Leverandør Toronto Research Chemicals Leverandørnr. TRCA192803100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 5-Acetyl-2H-pyrazole-3-carboxylic Acid Ethyl Ester
CAS 37622-89-2
Molekylær formel C8H10N2O3
Mængde 100 mg
InChI formel InChI=1S/C8H10N2O3/c1-3-13-8(12)7-4-6(5(2)11)9-10-7/h4H,3H2,1-2H3,(H,9,10)
SMIL CCOC(=O)c1cc(n[nH]1)C(=O)C
IUPAC navn ethyl 3-acetyl-1H-pyrazole-5-carboxylate
Molekylvægt (g/mol) 182.18
Formel vægt 182.0691
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.