Learn More
5-Acetyl-2-thiophenecarbaldehyde, 97%, Thermo Scientific™
Product Code. 10768423
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10768423 1 g 1g
10239352 250 mg 250mg
10002314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10768423 Supplier Thermo Scientific Maybridge Supplier No. MO01240DA
 Restricted Item
Forventet forsendelse: 01-12-2026
Add to basket
Chemical Identifiers
CAS 4565-29-1
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
InChI Key ISGDPVVLVWQENT-UHFFFAOYSA-N
Synonym 5-acetyl-2-thiophenecarbaldehyde, 2-acetyl-5-formylthiophene, 5-acetyl-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-acetyl
PubChem CID 2794713
IUPAC Name 5-acetylthiophene-2-carbaldehyde
SMILES CC(=O)C1=CC=C(S1)C=O
Specifications
Assay Percent Range 97%
Quantity 1 g
Formula Weight 154.19
Percent Purity 97%
Chemical Name or Material 5-Acetyl-2-thiophenecarbaldehyde