Få mere at vide
5-Acetyl-2-chloropyrazine, TRC
Artikelnummer. 30324235
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324235 500 mg
30316828 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30324235 Leverandør Toronto Research Chemicals Leverandørnr. TRCA172025500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Acetyl-2-chloropyrazine
CAS 160252-31-3
Molekylær formel C6 H5 Cl N2 O
Mængde 500 mg
Synonym 1-(5-Chloro-2-pyrazinyl)ethanone, 1-(5-Chloropyrazinyl)ethanone, 2-Chloro-5-acetylpyperazine
InChI formel InChI=1S/C6H5ClN2O/c1-4(10)5-2-9-6(7)3-8-5/h2-3H,1H3
SMIL CC(=O)c1cnc(Cl)cn1
IUPAC navn 1-(5-chloropyrazin-2-yl)ethanone
Molekylvægt (g/mol) 156.57
Formel vægt 156.009