missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Acetonyl-2-methoxybenzenesulfonamide, Mikromol™

Artikelnummer. 30463401
Change view
Click to view available options
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30463401 4 x 25 mg
30052554 100 mg
30051369 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463401 Supplier Mikromol Supplier No. MM0938.1200254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Acetonyl-2-methoxybenzenesulfonamide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Acetonyl-2-methoxybenzenesulfonamide
CAS 116091-63-5
Analyt- eller komponentnavne 5-Acetonyl-2-methoxybenzenesulfonamide
Forsendelsestilstand Room Temperature
Molekylær formel C10 H13 N O4 S
Certificeringer/compliance ISO 17025
Synonym 2-Methoxy-5-(2-oxopropyl)benzenesulfonamide, 5-Acetonyl-2-methoxybenzenesulfonamide, Benzenesulfonamide, 2-methoxy-5-(2-oxopropyl)-
InChI formel InChI=1S/C10H13NO4S/c1-7(12)5-8-3-4-9(15-2)10(6-8)16(11,13)14/h3-4,6H,5H2,1-2H3,(H2,11,13,14)
Løsningstype Neat
SMIL COc1ccc(CC(=O)C)cc1S(=O)(=O)N
IUPAC navn 2-methoxy-5-(2-oxopropyl)benzenesulfonamide
Molekylvægt (g/mol) 243.28
Mængde 4 x 25 mg
Formel vægt 243.0565 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.