Få mere at vide
5,8-Dioxo Psoralen, TRC
Artikelnummer. 30373639
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373639 25 mg
30373629 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30373639 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48685025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 483-36-3
Molekylær formel C11H4O5
Molekylvægt (g/mol) 216.15
Synonym 4H-Furo[3,2-g][1]benzopyran-4,7,9-trione, 4,7-Dihydro-6-hydroxy-4,7-dioxo-5-benzofuranacrylic Acid delta-Lactone, Bergaptenquinone, NSC 401268, Psoralen Quinone
SMIL O=C1OC2=C(C=C1)C(=O)c3ccoc3C2=O
Tekniske data
Kemisk navn eller materiale 5,8-Dioxo Psoralen
Mængde 25 mg
InChI formel InChI=1S/C11H4O5/c12-7-2-1-5-8(13)6-3-4-15-10(6)9(14)11(5)16-7/h1-4H
Formel vægt 216.0059