Få mere at vide
5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one, TRC
Artikelnummer. 30380948
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30380948 5 mg
30373836 50 mg
30375446 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30380948 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4532685MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 520-31-0
Molekylær formel C15H10O7
Molekylvægt (g/mol) 302.24
Synonym 5,7,3',4',5'-Pentahydroxyflavone, Tricetin
SMIL Oc1cc(O)c2C(=O)C=C(Oc2c1)c3cc(O)c(O)c(O)c3
Tekniske data
Kemisk navn eller materiale Tricetin
Mængde 5 mg
InChI formel InChI=1S/C15H10O7/c16-7-3-8(17)14-9(18)5-12(22-13(14)4-7)6-1-10(19)15(21)11(20)2-6/h1-5,16-17,19-21H
Formel vægt 302.0427