Få mere at vide
5,7-Dichloro-8-hydroxy-2-methylquinoline, TRC
Artikelnummer. 30136790
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136790 1 g
17953955 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136790 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4368231G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72-80-0
Molekylær formel C10 H7 Cl2 N O
Molekylvægt (g/mol) 228.07
Synonym 5,7-Dichloro-2-methyl-8-quinolinol, 5,7-Dichloro-2-methyl-8-hydroxyquinoline, 5,7-Dichloro-2-styrylquinoline, 5,7-Dichloro-8-hydroxy-2-methylquinoline, 5,7-Dichloro-8-hydroxyquinaldine, 5,7-Dichloro-8-quinaldinol, Acnosan, Afungil, Chloquinan, Chlorchinaldin, Chlorchinaldol, Chlorguinaldon, Chlorquinaldol, Florabina, Gyno-Sterosan, Gynotherax, Hydroxydichloroquinaldinol, Intensol, Quesil, Saprosan, Siogen, Siogenal, Siogene, Siogeno, Siogenon, Siosept, Siosteran, Sterosan, Steroxin, Sterozan, Vagisteran
IUPAC navn 5,7-dichloro-2-methylquinolin-8-ol
SMIL Cc1ccc2c(Cl)cc(Cl)c(O)c2n1
Tekniske data
Kemisk navn eller materiale Chlorquinaldol
Mængde 1 g
InChI formel InChI=1S/C10H7Cl2NO/c1-5-2-3-6-7(11)4-8(12)10(14)9(6)13-5/h2-4,14H,1H3
Formel vægt 226.9905