Få mere at vide
5,6-trans-Vitamin D2, TRC
Artikelnummer. 30455320
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30455320 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455320 Leverandør Toronto Research Chemicals Leverandørnr. TRCV67606050MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5,6-trans-Vitamin D2
CAS 51744-66-2
Molekylær formel C28H44O
Mængde 50 mg
Synonym 9,10-Secoergosta-5,7,10(19),22-tetraen-3-ol','(3β,5E,7E,22E)- (9CI), (5E,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraen-3β-ol, (5E)-Ergocalciferol, 5,6-trans-Ergocalciferol, 5,6-trans-Vitamin D2
InChI formel InChI=1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13+/t20-,22+,25-,26+,27-,28+/m0/s1
SMIL CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C\3/C[C@@H](O)CCC3=C)\CCC[C@]12C
IUPAC navn (1S,3E)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molekylvægt (g/mol) 396.65
Formel vægt 396.3392