Få mere at vide
5,6-Dimethylbenzotriazole Hydrate, TRC
Artikelnummer. 17901725
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17901725 5 g
17972165 1 g
17920595 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17901725 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4622755G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1354973-50-4
Molekylær formel C8 H9 N3 . H2 O
Molekylvægt (g/mol) 165.19
Synonym 5,6-Dimethyl-1H-benzotriazole monohydrate, 5,6-Dimethyl-1,2,3-benzotriazole Hydrate, 5,6-Dimethylbenzotriazole Hydrate, NSC 62005 Hydrate
IUPAC navn 5,6-dimethyl-1H-benzotriazole;hydrate
SMIL O.Cc1cc2nn[nH]c2cc1C
Tekniske data
Kemisk navn eller materiale 5,6-Dimethyl-1H-benzotriazole monohydrate
Mængde 5 g
InChI formel InChI=1S/C8H9N3.H2O/c1-5-3-7-8(4-6(5)2)10-11-9-7;/h3-4H,1-2H3,(H,9,10,11);1H2
Formel vægt 165.0902