missing translation for 'onlineSavingsMsg'
Få mere at vide

5,6-Dimethoxy-2-(4-pyridylmethylene)-1-indanone (Donepezil Impurity), TRC

Artikelnummer. 30387895
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30387895 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30387895 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4607202.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 5,6-Dimethoxy-2-[(4-pyridyl)methylene]indan-1-one
CAS 4803-74-1
Molekylær formel C17 H15 N O3
Mængde 2.5 g
Synonym 5,6-Dimethoxy-2-[(4-pyridyl)methylene]indan-1-one
InChI formel InChI=1S/C17H15NO3/c1-20-15-9-12-8-13(7-11-3-5-18-6-4-11)17(19)14(12)10-16(15)21-2/h3-7,9-10H,8H2,1-2H3/b13-7-
SMIL COc1cc2C\C(=C\c3ccncc3)\C(=O)c2cc1OC
IUPAC navn (2Z)-5,6-dimethoxy-2-(pyridin-4-ylmethylidene)-3H-inden-1-one
Molekylvægt (g/mol) 281.31
Formel vægt 281.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.