Få mere at vide
5,6-Dimethoxy-2-[(4-pyridyl)methylene]indan-1-one, Mikromol™
Artikelnummer. 30050423
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050423 100 mg
30050110 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050423 Leverandør Mikromol Leverandørnr. MM0677.13
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5,6-Dimethoxy-2-[(4-pyridyl)methylene]indan-1-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5,6-Dimethoxy-2-[(4-pyridyl)methylene]indan-1-one
CAS 4803-74-1
Analyt- eller komponentnavne 5,6-Dimethoxy-2-[(4-pyridyl)methylene]indan-1-one
Forsendelsestilstand Room Temperature
Molekylær formel C17 H15 N O3
Certificeringer/compliance ISO 17025
Synonym 5,6-Dimethoxy-2-[(4-pyridyl)methylene]indan-1-one
InChI formel InChI=1S/C17H15NO3/c1-20-15-9-12-8-13(7-11-3-5-18-6-4-11)17(19)14(12)10-16(15)21-2/h3-7,9-10H,8H2,1-2H3/b13-7-
SMIL COc1cc2C\C(=C\c3ccncc3)\C(=O)c2cc1OC
IUPAC navn (2Z)-5,6-dimethoxy-2-(pyridin-4-ylmethylidene)-3H-inden-1-one
Molekylvægt (g/mol) 281.31
Mængde 100 mg
Formel vægt 281.1052 g/mol