missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydroxyflavone, 97%, Thermo Scientific Chemicals

Catalog Number p-7029563
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
6665-66-3
C15H10O4
254.241
MFCD00601534
AGZAGADSYIYYCT-UHFFFAOYSA-N
5,6-dihydroxyflavone, 5,6-dihydroxy-2-phenyl-4h-chromen-4-one, 5,6-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 5,6-dihydroxy-2-phenyl
14349487
5,6-dihydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC(=C3O)O
This item is not returnable. View return policy
6665-66-3
C15H10O4
254.241
MFCD00601534
AGZAGADSYIYYCT-UHFFFAOYSA-N
5,6-dihydroxyflavone, 5,6-dihydroxy-2-phenyl-4h-chromen-4-one, 5,6-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 5,6-dihydroxy-2-phenyl
14349487
5,6-dihydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC(=C3O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 254.24
Percent Purity 97%
Chemical Name or Material 5,6-Dihydroxyflavone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'