Få mere at vide
5,6-Dihydrouracil-5,5,6,6-d4, CDN
Artikelnummer. 30321296
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321296 0.005 g
30308951 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321296 Leverandør CDN Leverandørnr. CDND76880.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dihydro Uracil-d4
CAS 334473-41-5
Molekylær formel C4 D4 H2 N2 O2
Mængde 0.005 g
Synonym 2,4(1 H,3 H)-Pyrimidinedione-5,6-d2, dihydro-d2- (9 CI), Dihydro-d2-2,4(1 H,3 H)-pyrimidinedione-5,6-d2, Hydrouracil-d4, 5,6-Dihydro-2,4(1 H,3 H)-pyrimidinedione-5,5,6,6-d4, 5,6-Dihydro-2,4-dihydroxypyrimidine-5,5,6,6-d4, 5,6-Dihydrouracil-d4, 5,6-Dihydrouracil-5,5,6,6-D4, Dihydrouracil-d4
InChI formel InChI=1 S/C4H6N2O2/c7-3-1-2-5-4(8)6-3/h1-2H2,(H2,5,6,7,8)/i1D2,2D2
SMIL [2 H]C1([2 H])NC(=O)NC(=O)C1([2 H])[2 H]
IUPAC navn 5,5,6,6-tetradeuterio-1,3-diazinane-2,4-dione
Molekylvægt (g/mol) 118.127
Formel vægt 118.068 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity