Få mere at vide
5,6-Dihydro Thymine-d6, TRC
Artikelnummer. 30377063
Skift visning
Mængde:
1 mg
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30377063 2.5 mg 2.5mg
30368794 25 mg -
30370710 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30377063 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4494422.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 334473-42-6
Molekylær formel C5 2H6 H2 N2 O2
Molekylvægt (g/mol) 134.17
Synonym 2,4(1H,3H)-Pyrimidinedione-5,6-d2, dihydro-6-d-5-(methyl-d3)- (9CI), 5-Methylhydrouracil-d6, Dihydro-5-(methyl-d3)-2,4(1H,3H)-pyrimidinedione-5,6,6-d3, 5,6-Dihydro-5-methyluracil-α,α,α,5,6,6-D6, 5,6-Dihydrothymine-d6, 5-(Methyl-d3)-5,6,6-d3-5,6-dihydrouracil, 5-(Methyl-d3)dihydropyrimidine-2,4(1H,3H)-dione-5,6,6-d3, Dihydrothymine-d6, Dihydrothymine-α,α,α,5,6,6-d6
IUPAC navn 5,6,6-trideuterio-5-(trideuteriomethyl)-1,3-diazinane-2,4-dione
SMIL [2H]C([2H])([2H])C1([2H])C(=O)NC(=O)NC1([2H])[2H]
Tekniske data
Kemisk navn eller materiale 5,6-Dihydro-5-methyluracil alpha,alpha,alpha,5,6,6-d6
Mængde 2.5 mg
InChI formel InChI=1S/C5H8N2O2/c1-3-2-6-5(9)7-4(3)8/h3H,2H2,1H3,(H2,6,7,8,9)/i1D3,2D2,3D
Formel vægt 134.0962