missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dicyano-3-thienyl-2-pyrazinone, 97%, Thermo Scientific™

Catalog Number p-7039287
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1421263-07-1
C10H4N4OS
228.229
MFCD22988953
UNKZQYRUAFQDJO-UHFFFAOYSA-N
5,6-dicyano-3-thienyl-2-pyrazinone, 5-oxo-6-thiophen-3-yl-4h-pyrazine-2,3-dicarbonitrile
97030351
6-oxo-5-thiophen-3-yl-1H-pyrazine-2,3-dicarbonitrile
C1=CSC=C1C2=NC(=C(NC2=O)C#N)C#N
This item is not returnable. View return policy
1421263-07-1
C10H4N4OS
228.229
MFCD22988953
UNKZQYRUAFQDJO-UHFFFAOYSA-N
5,6-dicyano-3-thienyl-2-pyrazinone, 5-oxo-6-thiophen-3-yl-4h-pyrazine-2,3-dicarbonitrile
97030351
6-oxo-5-thiophen-3-yl-1H-pyrazine-2,3-dicarbonitrile
C1=CSC=C1C2=NC(=C(NC2=O)C#N)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 228.23
Percent Purity 97%
Chemical Name or Material 5,6-Dicyano-3-thienyl-2-pyrazinone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'