missing translation for 'onlineSavingsMsg'
Få mere at vide

5,6-Dichloropurine-1-(2,3,5-tri-O-acetyl-Beta-D-ribofuanosyl)-1H-benzimidazole, TRC

Artikelnummer. 30369872
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30369872 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30369872 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43593550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 5,6-Dichloropurine-1-(2,3,5-tri-O-acetyl-beta-D-ribofuanosyl)-1H-benzimidazole
CAS 443678-71-5
Molekylær formel C18H18Cl2N2O7
Mængde 50 mg
Synonym 5,6-Dichloro-1-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)-1H-benzimidazole
InChI formel InChI=1S/C18H18Cl2N2O7/c1-8(23)26-6-15-16(27-9(2)24)17(28-10(3)25)18(29-15)22-7-21-13-4-11(19)12(20)5-14(13)22/h4-5,7,15-18H,6H2,1-3H3/t15-,16-,17-,18-/m1/s1
SMIL CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)n2cnc3cc(Cl)c(Cl)cc23
IUPAC navn [(2R,3R,4R,5R)-3,4-diacetyloxy-5-(5,6-dichlorobenzimidazol-1-yl)oxolan-2-yl]methyl acetate
Molekylvægt (g/mol) 445.25
Formel vægt 444.0491
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.