missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dichlorobenzimidazole riboside, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Description
Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.5,6-Dichlorobenzimidazole riboside is used as an inhibitor of RNA Polymerase II. It is nucleoside analogue and used in research to study the mechanisms of cellular regulation. Further, it inhibits some protein kinases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.5,6-Dichlorobenzimidazole riboside is used as an inhibitor of RNA Polymerase II. It is nucleoside analogue and used in research to study the mechanisms of cellular regulation. Further, it inhibits some protein kinases.
Solubility
Soluble in DMSO to 75mMSoluble in dimethyl sulfoxide, hot ethanol, dimethyl formamide, (1:2) dimethyl formamide:Phosphate buffer saline, water, methanol and pyridine. Slightly soluble in aqueous buffers.
Notes
Incompatible with heat and oxidizing agents.
Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.5,6-Dichlorobenzimidazole riboside is used as an inhibitor of RNA Polymerase II. It is nucleoside analogue and used in research to study the mechanisms of cellular regulation. Further, it inhibits some protein kinases.
Solubility
Soluble in DMSO to 75mMSoluble in dimethyl sulfoxide, hot ethanol, dimethyl formamide, (1:2) dimethyl formamide:Phosphate buffer saline, water, methanol and pyridine. Slightly soluble in aqueous buffers.
Notes
Incompatible with heat and oxidizing agents.
Specifications
Color | White |
Quantity | 100 mg |
Beilstein | 39123 |
Solubility Information | Soluble in DMSO to 75mMSoluble in dimethyl sulfoxide,hot ethanol,dimethyl formamide,(1:2) dimethyl formamide:Phosphate buffer saline,water,methanol and pyridine; Slightly soluble in aqueous buffers. |
Formula Weight | 319.14 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 5,6-Dichlorobenzimidazole riboside |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'