Få mere at vide
5,6-Dibromo-1,10,11,12,13,13-hexachloro-11-tricyclo[8.2.1.02,9]tridecene, TRC
Artikelnummer. 30007938
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007938 10 mg
30006694 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007938 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41873510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51936-55-1
Molekylær formel C13 H12 Br2 Cl6
Molekylvægt (g/mol) 540.76
Synonym 1,4-Methanobenzocyclooctene, 7,8-dibromo-1,2,3,4,11,11-hexachloro-1,4,4a,5,6,7,8,9,10,10a-decahydro-, 7,8-Dibromo-1,2,3,4,11,11-hexachloro-1,4,4a,5,6,7,8,9,10,10a-decahydro-1,4-methanobenzocyclooctene, 5,6-Dibromo-1,10,11,12,13,13-hexachloro-11-tricyclo[8.2.1.02,9]tridecene, Citex BC 26, HCDBCO, Saytex BC 26
SMIL ClC1=C(Cl)C2(Cl)C3CCC(Br)C(Br)CCC3C1(Cl)C2(Cl)Cl
Tekniske data
Kemisk navn eller materiale Hexachlorocyclopentadienyldibromocyclooctane
Mængde 10 mg
InChI formel InChI=1S/C13H12Br2Cl6/c14-7-3-1-5-6(2-4-8(7)15)12(19)10(17)9(16)11(5,18)13(12,20)21/h5-8H,1-4H2
Formel vægt 535.7437