Få mere at vide
5,6,7,8-Tetrahydroisoquinolin-1-amine, TRC
Artikelnummer. 30418926
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418926 10 mg
30416028 100 mg
30415544 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30418926 Leverandør Toronto Research Chemicals Leverandørnr. TRCT20561810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75704-51-7
Molekylær formel C9 H12 N2
Molekylvægt (g/mol) 148.21
IUPAC navn 5,6,7,8-tetrahydro-2H-isoquinolin-1-imine
SMIL N=C1NC=CC2=C1CCCC2
Tekniske data
Kemisk navn eller materiale 5,6,7,8-Tetrahydroisoquinolin-1-amine
Mængde 10 mg
InChI formel InChI=1S/C9H12N2/c10-9-8-4-2-1-3-7(8)5-6-11-9/h5-6H,1-4H2,(H2,10,11)
Formel vægt 148.1